Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neuroligin 1/NLGN1 Antibody (S97A-31), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242192IR
Description
Neuroligin 1/NLGN1 Monoclonal specifically detects Neuroligin 1/NLGN1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Neuroligin 1/NLGN1 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
NLGN1 | |
Fusion protein amino acids 718-843 (cytoplasmic C-terminus) of rat Neuroligin-1. Mouse: 99% identity (125/126 amino acids identical). Human: 99% identity (125/126 amino acids identical) >40% identity with Neuroligin-2 and -3. | |
0.1 mL | |
Primary | |
Greater then 40% identity with Neuroligin-2 and -3.Detects approx 120kDa. Does not cross-react with other Neuroligins. | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
S97A-31 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
EC 3.1.1, EC 3.1.1.1, KIAA1070neuroligin-1, MGC45115, neuroligin 1, NL1 | |
Mouse | |
Protein G purified | |
Cognition and Behavior | |
22871 | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction