Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Neuroligin 1 Recombinant Rabbit Monoclonal Antibody (001), Invitrogen™
GREENER_CHOICE

Catalog No. PIMA543128
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543128 100 μL
PIMA543433 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543128 Supplier Invitrogen™ Supplier No. MA543128
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Specificity: Human Neuroligin 1/NLGN1

This gene encodes a member of a family of neuronal cell surface proteins. Members of this family may act as splice site-specific ligands for beta-neurexins and may be involved in the formation and remodeling of central nervous system synapses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neuroligin 1
Applications ELISA
Classification Recombinant Monoclonal
Clone 001
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Nlgn1
Gene Accession No. Q8N2Q7
Gene Alias 6330415N05Rik; BB179718; Kiaa1070; MGC45115; mKIAA1070; neuroligin 1; neuroligin I; Neuroligin-1; NL1; NLG 1; Nlg1; NLGN1
Gene Symbols Nlgn1
Host Species Rabbit
Immunogen Recombinant Human Neuroligin 1/NLGN1 protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22871
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.