Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neuroligin 2/NLGN2 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP100220
Description
Neuroligin 2/NLGN2 Polyclonal specifically detects Neuroligin 2/NLGN2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Peptide ELISA.Specifications
Neuroligin 2/NLGN2 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
EC 3.1.1, EC 3.1.1.1, KIAA1366neuroligin-2, neuroligin 2 | |
Goat | |
91 kDa | |
0.1 mg | |
Cognition and Behavior | |
57555 | |
Human | |
IgG |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, ELISA | |
0.5 mg/mL | |
Western Blot 2 - 3 ug/mL, Flow Cytometry 10 ug/mL, Immunocytochemistry/Immunofluorescence 10 ug/mL, Peptide ELISA Detection limit 1:4000 | |
Q8NFZ4 | |
NLGN2 | |
Peptide with sequence C-NPPDTDIRDPGKK corresponding to internal region according to NP_065846.1. | |
Affinity Purified | |
RUO | |
Primary | |
Human and Rat. Expected from sequence similarity: Mouse and Dog. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction