Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Neuroligin 2/NLGN2 Antibody, PerCP, Novus Biologicals™
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241299PCP
Description
Neuroligin 2/NLGN2 Polyclonal antibody specifically detects Neuroligin 2/NLGN2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Neuroligin 2/NLGN2 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| EC 3.1.1, EC 3.1.1.1, KIAA1366neuroligin-2, neuroligin 2 | |
| Antibody was raised against a 19 amino acid peptide near the carboxy terminus of human NLGN2. The immunogen is located within amino acids 630 - 680 of NLGN2. Amino Acid Squence: PPATLPPEPEPEPGPRAYD | |
| 0.1 mL | |
| Cognition and Behavior | |
| 57555 | |
| Store at 4C in the dark. | |
| IgG | 
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| PerCP | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction