Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Neuroligin 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564404
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564404 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564404 Supplier Invitrogen™ Supplier No. PA564404
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSLHPFGPFP PPPPTATSHN NTLPH Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 88%.

This gene encodes a member of a family of neuronal cell surface proteins. Members of this family may act as splice site-specific ligands for beta-neurexins and may be involved in the formation and remodeling of central nervous system synapses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neuroligin 2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NLGN2
Gene Accession No. Q8NFZ4
Gene Alias Kiaa1366; neuroligin 2; neuroligin-2; NL2; Nlgn2; ortholog of human and rat neuroligin 2 NLGN2
Gene Symbols NLGN2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NLGN2. Recombinant protein control fragment (Product #RP-103833).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57555
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.