Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Neuroligin 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589727
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589727 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589727 Supplier Invitrogen™ Supplier No. PA589727
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QRGGGGPGGG APGGPGLGLG SLGEERFPVV NTAYGRVRGV RRELNNEILG PVVQFLGVPY ATPPLGARRF QPPEAPASWP GVRNATTLPP ACPQNLHGAL PAIMLPVWFT DNLEAAATYV QNQSEDCLYL NLYVPTEDGP LTKKRDEATL NPPDTDIRDP GKKPVM; Positive Samples: Mouse heart; Cellular Location: Cell junction, Cell membrane, Single-pass type I membrane protein, postsynaptic cell membrane, presynaptic cell membrane, synapse.

This gene encodes a member of a family of neuronal cell surface proteins. Members of this family may act as splice site-specific ligands for beta-neurexins and may be involved in the formation and remodeling of central nervous system synapses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neuroligin 2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NLGN2
Gene Accession No. Q69ZK9
Gene Alias Kiaa1366; neuroligin 2; neuroligin-2; NL2; Nlgn2; ortholog of human and rat neuroligin 2 NLGN2
Gene Symbols NLGN2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 15-180 of human NLGN2 (NP_0658461).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216856
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.