Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuroligin-3 Monoclonal Antibody (S110-29), QED Bioscience™
SDP

Catalog No. 502278237
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-8237 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-8237 Supplier Leinco Technologies Inc Supplier No. 56552100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Neuroligin-3 Monoclonal antibody specifically detects Neuroligin-3 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen Neuroligin-3
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone S110-29
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.4
Gene NLGN3
Gene Accession No. Q62889, Q8BYM5, Q9NZ94
Gene Alias A230085M13Rik; ASPGX1; AUTSX1; Gliotactin homolog; HNL3; KIAA1480; neuroligin 3; neuroligin 3 protein; neuroligin-3; NL3; NLGN3
Host Species Mouse
Immunogen Fusion protein corresponding to aa 730-848 (intracellular C-terminus) of rat Neuroligin-3. This sequence is 99
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171297, 245537, 54413
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.