Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neuroligin 4X/NLGN4X Antibody (S98-7), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242198H
Description
Neuroligin 4X/NLGN4X Monoclonal specifically detects Neuroligin 4X/NLGN4X in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Neuroligin 4X/NLGN4X | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
NLGN4X | |
Fusion protein amino acids 782-945 (intracellular C-terminus) of mouse Neuroligin 4. ∼45% identity with Neuroligin-1, -2 and -3. | |
0.1 mL | |
57502 | |
Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
S98-7 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
ASPGX2, HLNX, HNLX, KIAA1260AUTSX2, MGC22376, neuroligin 4, X-linked, NLGN, NLGN4neuroligin 4, X-linked | |
Mouse | |
Protein G purified | |
Primary | |
Approx 45% identity with Neuroligin-1, -2 and -3.Detects approx 25-130kDa. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction