Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neurolysin Antibody (OTI1D6), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272961MFV610
Description
Neurolysin Monoclonal antibody specifically detects Neurolysin in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Flow CytometrySpecifications
Neurolysin | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Primate, Monkey | |
Purified |
Flow Cytometry | |
OTI1D6 | |
Flow Cytometry | |
AGTBPFLJ23002, angiotensin binding protein, Angiotensin-binding protein, DKFZp564F123, EC 3.4.24, EC 3.4.24.16, KIAA1226EP24.16, MEP, mitochondrial, MOP, neurolysin (metallopeptidase M3 family) | |
Full length human recombinant protein of human NLN(NP_065777) produced in HEK293T cell. | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
57486 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction