Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neurolysin Antibody (OTI1D6), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272961PE
Description
Neurolysin Monoclonal specifically detects Neurolysin in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Flow Cytometry.Specifications
Neurolysin | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
NLN | |
Full length recombinant protein of human Neurolysin (NP_065777) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Primate, Monkey | |
IgG1 |
Flow Cytometry | |
OTI1D6 | |
Flow Cytometry | |
AGTBPFLJ23002, angiotensin binding protein, Angiotensin-binding protein, DKFZp564F123, EC 3.4.24, EC 3.4.24.16, KIAA1226EP24.16, MEP, mitochondrial, MOP, neurolysin (metallopeptidase M3 family) | |
Mouse | |
Affinity Purified | |
Core ESC Like Genes, Stem Cell Markers | |
57486.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction