Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neuromedin BR/NMBR Antibody (466505), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4728E
Description
Neuromedin BR/NMBR Monoclonal antibody specifically detects Neuromedin BR/NMBR in Human samples. It is validated for Flow Cytometry, Flow Cytometry (Intracellular Staining), CyTOFSpecifications
Neuromedin BR/NMBR | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Flow Cytometry (Intracellular Staining), CyTOF | |
466505 | |
Flow Cytometry, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
neuromedin B receptor, neuromedin-B receptor, Neuromedin-B-preferring bombesin receptor, NMB-R | |
NS0 mouse myeloma cell line transfected with human NMBR, Met1-Met390, Accession # AAB27330 | |
0.1 mL | |
GPCR | |
4829 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction