Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Neuromedin U Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596566
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596566 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596566 Supplier Invitrogen™ Supplier No. PA596566
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive samples: Mouse large intestine, Mouse stomach, Rat large intestine, Rat stomach; Cellular location: Secreted Immunogen sequence: ACRGAPILPQ GLQPEQQLQL WNEIDDTCSS FLSIDSQPQA SNALEELCFM IMGMLPKPQE QDEKDNTKRF LFHYSKTQKL GKSNVVSSVV HPLLQLVPHL HERRMKRFRV DEEFQSPFAS QSRGYFLFRP RNGRRSAGFI.

This gene encodes a member of the neuromedin family of neuropeptides. The encoded protein is a precursor that is proteolytically processed to generate a biologically active neuropeptide that plays a role in pain, stress, immune-mediated inflammatory diseases and feeding regulation. Increased expression of this gene was observed in renal, pancreatic and lung cancers. Alternative splicing results in multiple transcript variants encoding different isoforms. Some of these isoforms may undergo similar processing to generate the mature peptide.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neuromedin U
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.86 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene NMU
Gene Accession No. P12760, P48645, Q9QXK8
Gene Alias Neuromedin precursor-related peptide 33; Neuromedin precursor-related peptide 36; neuromedin U; neuromedin-U; Neuromedin-U-23; Neuromedin-U-25; Nmu; NmU 25; NmU-23; NmU-25; NURP33; NURP36; prepro-NMU
Gene Symbols NMU
Host Species Rabbit
Immunogen Synthetic peptide
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10874, 56183, 63887
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.