Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuron-specific beta-III Tubulin APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC1195A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC1195A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC1195A Supplier R&D Systems Supplier No. IC1195A
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

beta-III Tubulin Monoclonal specifically detects beta-III Tubulin in Multi-Species samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen beta-III Tubulin
Applications Flow Cytometry
Classification Monoclonal
Clone TuJ-1
Conjugate APC
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Alias Beta3-tubulin, beta-4, betaIII Tubulin, beta-Tubulin III, Class III beta-tubulin, TUBB3, TUBB4 CFEOM3A, Tubulin Beta 3, tubulin beta-3 chain, Tubulin beta-4, Tubulin beta-4 chain, Tubulin beta-III, tubulin, beta 3, TUJ1 antigen, TUJ-1 antigen
Gene Symbols TUBB3
Host Species Mouse
Immunogen Rat brain-derived microtubules
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10381
Test Specificity Detects mammalian and chicken neuron-specific beta-III tubulin but not other beta-tubulin isotypes in flow cytometry.
Target Species Multi-species
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.