Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuron-specific beta-III Tubulin NorthernLights™ NL557-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. NL1195R
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NL1195R 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NL1195R Supplier R&D Systems Supplier No. NL1195R
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 6 publications

beta-III Tubulin Monoclonal specifically detects beta-III Tubulin in Multi-Species samples. It is validated for Immunocytochemistry.

Specifications

Antigen beta-III Tubulin
Applications Immunocytochemistry
Classification Monoclonal
Clone TuJ-1
Conjugate NorthernLights 557 (NL557)
Dilution Immunocytochemistry 1:10 dilution
Formulation Supplied as a 10X solution of antibody in PBS containing sodium azide. with Sodium Azide
Gene Alias Beta3-tubulin, beta-4, betaIII Tubulin, beta-Tubulin III, Class III beta-tubulin, TUBB3, TUBB4 CFEOM3A, Tubulin Beta 3, tubulin beta-3 chain, Tubulin beta-4, Tubulin beta-4 chain, Tubulin beta-III, tubulin, beta 3, TUJ1 antigen, TUJ-1 antigen
Gene Symbols TUBB3
Host Species Mouse
Immunogen Rat brain-derived microtubules
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10381
Test Specificity Detects mammalian and chicken neuron-specific beta-III tubulin but not other beta-tubulin isotypes in Western blots.
Target Species Multi-species
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.