Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neuronal Pentraxin 1 Antibody (OTI5G9), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272965C
Description
Neuronal Pentraxin 1 Monoclonal specifically detects Neuronal Pentraxin 1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
| Neuronal Pentraxin 1 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| NPTX1 | |
| Full length recombinant protein of human Neuronal Pentraxin 1 (NP_002513) produced in HEK293T cells. | |
| 0.1 mL | |
| 4884.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI5G9 | |
| Western Blot | |
| DKFZp686J2446, MGC105123, neuronal pentraxin INP1NP-I, neuronal pentraxin-1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction