Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neuronal Pentraxin 2 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176899PCP
Description
Neuronal Pentraxin 2 Polyclonal antibody specifically detects Neuronal Pentraxin 2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Neuronal Pentraxin 2 | |
Polyclonal | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
NARP, neuronal activity-regulated pentaxin, neuronal pentraxin IIapexin, neuronal pentraxin-2, NP2, NP-II | |
Antibody was raised against a 16 amino acid synthetic peptide near the center of the human NPTX2. The immunogen is located within amino acids 170 - 220 of NPTX2. Amino Acid Squence: LHNETSAHRQKTESTL | |
0.1 mL | |
Stem Cell Markers | |
4885 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction