Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neuronal Pentraxin R/NPTXR Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310142100UL
Description
Neuronal Pentraxin R/NPTXR Polyclonal specifically detects Neuronal Pentraxin R/NPTXR in Mouse samples. It is validated for Western Blot.Specifications
Neuronal Pentraxin R/NPTXR | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
neuronal pentraxin receptor, NPR | |
The immunogen is a synthetic peptide directed towards the middle region of mouse Neuronal Pentraxin R/NPTXR. Peptide sequence VAQLPLSLKDSNWHHICISWTTRDGLWSAYQDGELRGSGENLAAWHPIKP | |
100 μg | |
Primary | |
Mouse | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
23467 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction