Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Neuropeptide B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120468
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120468 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120468 Supplier Invitrogen™ Supplier No. PA5120468
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: SW480, Mouse brain. The target is usually found in the following locations: Secreted. Immunogen sequence: MARSATLAAA ALALCLLLAP PGLAWYKPAA GHSSYSVGRA AGLLSGLRRS PYARRSQPYR GAEPPGGAGA SPELQLHPRL RSLAVCVQDV APNLQRCERL PDGRGTYQCK ANVFLSLRAA DCLAA.

Neuropeptide B is a endrogenous ligand of the orphan G protein coupled receptor GPR7. NPB and GPR7 exist in close proximity in the rat hypothalamus, and are ideally positioned to modulate neuroendocrine functions. NPB-immunoreactivity was enriched in many regions within the hypothalamus which also contained high levels of GPR7 mRNA including the ventrormedial hypothalamic nucleus, dorsomedial hypothalamic nucleus, arcuate nucleus, supraoptic retrochiasmatic nucleus, and in the area ventral to the zona incerta.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neuropeptide B
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NPB
Gene Accession No. Q8K4P1, Q8NG41
Gene Alias hPPL7; L7; L7C; mPPL7; neuropeptide B; Neuropeptide B-23; Neuropeptide B-29; NPB; NPB23; NPB29; PPL7; PPNPB; preproneuropeptide B; prepro-NPB; preproprotein L7; rPPL7
Gene Symbols NPB
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-125 of human NPB (NP_683694.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 208990, 256933
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.