Learn More
Invitrogen™ Neuropeptide B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA3303
Description
PA3-303 detects neuropeptide B from human, mouse and rat samples. PA3-303 has been successfully used in Western blot and immunofluorescence procedures. The PA3-303 immunogen is a synthetic peptide corresponding to residues W(1) Y K P A A G P H H Y S V G R A S G L L S S F H R F P S T(29) of mouse NPB protein.
Neuropeptide B is a endrogenous ligand of the orphan G protein coupled receptor GPR7. NPB and GPR7 exist in close proximity in the rat hypothalamus, and are ideally positioned to modulate neuroendocrine functions. NPB-immunoreactivity was enriched in many regions within the hypothalamus which also contained high levels of GPR7 mRNA including the ventrormedial hypothalamic nucleus, dorsomedial hypothalamic nucleus, arcuate nucleus, supraoptic retrochiasmatic nucleus, and in the area ventral to the zona incerta.
Specifications
| Neuropeptide B | |
| Polyclonal | |
| Unconjugated | |
| NPB | |
| hPPL7; L7; L7C; mPPL7; neuropeptide B; Neuropeptide B-23; Neuropeptide B-29; NPB; NPB23; NPB29; PPL7; PPNPB; preproneuropeptide B; prepro-NPB; preproprotein L7; rPPL7 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry, Western Blot | |
| Conc. Not Determined | |
| whole serum with 0.05% sodium azide | |
| Q8K4P1, Q8K4P2, Q8NG41 | |
| NPB | |
| Synthetic peptide corresponding to residues W(1) Y K P A A G P H H Y S V G R A S G L L S S F H R F P S T(29) of mouse NPB protein. | |
| RUO | |
| 208990, 256933, 259222 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.