Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neuropeptide FF R1/NPFFR1 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB300168F
Description
Neuropeptide FF R1/NPFFR1 Polyclonal antibody specifically detects Neuropeptide FF R1/NPFFR1 in Human samples. It is validated for Western BlotSpecifications
| Neuropeptide FF R1/NPFFR1 | |
| Polyclonal | |
| Western Blot | |
| FLJ10751, G protein-coupled receptor 147, GPR147, neuropeptide FF 1, neuropeptide FF receptor 1, NPFF1G-protein coupled receptor 147, NPFF1R1, OT7T022, RFamide-related peptide receptor OT7T022 | |
| N terminal sequence MEGEPSQPPNSSWPLS and the C terminal sequence CSHLPLTIPAWDI of the human NPFF1 protein. | |
| 0.1 mL | |
| GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 64106 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| FITC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction