Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Neuropeptide Y Receptor 6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113311
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113311 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113311 Supplier Invitrogen™ Supplier No. PA5113311
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Neuropeptide Y Receptor 6 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Neuropeptide Y Receptor Y6 (NPY6R) is a receptor for neuropeptide Y and peptide YY. The activity of this receptor is mediated by G proteins that inhibits adenylate cyclase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neuropeptide Y Receptor 6
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene NPY6R
Gene Accession No. Q61212, Q99463
Gene Alias neuropeptide Y receptor type 6; neuropeptide Y receptor Y6; neuropeptide Y receptor Y6 (pseudogene); NPY1RL; Npy5r; NPY6R; NPY6-R; NPY6RP; pancreatic polypeptide receptor 2; PP2; Ppyr2; Y2B
Gene Symbols NPY6R
Host Species Rabbit
Immunogen Recombinant Human Putative neuropeptide Y receptor type 6 protein (1-39AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18169, 4888
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.