Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neuropilin-1 Antibody (446921), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB3870J
Description
Neuropilin-1 Monoclonal antibody specifically detects Neuropilin-1 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
Neuropilin-1 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
446921 | |
Flow Cytometry, CyTOF-ready | |
BDCA4, CD304, CD304 antigen, DKFZp686A03134, DKFZp781F1414, neuropilin 1, neuropilin-1, NP1, NRP, transmembrane receptor, Vascular endothelial cell growth factor 165 receptor, VEGF165R | |
Mouse myeloma cell line NS0-derived recombinant human Neuropilin-1, Phe22-Lys644, Accession # NP_001019799 | |
0.1 mL | |
Adaptive Immunity, Angiogenesis, Breast Cancer, Cancer, Cardiovascular Biology, Neuroscience, Virology Bacteria and Parasites | |
8829 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction