Learn More
Neuropilin-1 (CD304) Mouse anti-Human, BB515, Clone: U21-1283, BD Horizon™
Shop All BD Biosciences ProductsDescription
Specifications
Specifications
| Antigen | Neuropilin-1 (CD304) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | U21-1283 |
| Conjugate | Brilliant Blue 515 (BB515) |
| Description | Vol. Per test: 5μL |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | Neuropilin; NRP; NP1; Blood dendritic cell antigen 4; BDCA-4; VEGF165R |
| Host Species | Mouse |
| Immunogen | Recombinant Human Neuropilin-1 |
| Show More |
For Reseach Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.