Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neuropilin-1 (CD304) Mouse anti-Human, PE, Clone: U21-1283, BD Pharmingen™
Mouse Monoclonal Antibody
$164.70 - $361.50
Specifications
Antigen | Neuropilin-1 (CD304) |
---|---|
Clone | U21-1283 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE |
Description
Neuropilin-1 (CD304) Monoclonal antibody specifically detects Neuropilin-1 (CD304) in Human samples. It is validated for Flow cytometry.
Neuropilin-1 (CD304) Monoclonal antibody specifically detects Neuropilin-1 (CD304) in Human samples. It is validated for Flow cytometry.Specifications
Neuropilin-1 (CD304) | |
Flow Cytometry | |
PE | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Recombinant Human Neuropilin-1 | |
Primary |
U21-1283 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Neuropilin; NRP; NP1; Blood dendritic cell antigen 4; BDCA-4; VEGF165R | |
IgG2b κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions and unconjugated antibody and free PE were removed. |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title