Learn More
Invitrogen™ Neuropilin 1 Monoclonal Antibody (8G6G7)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA549195
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Human Neuropilin 1 shares 95% and 94% amino acid (aa) sequences identity with mouse and rat Neuropilin 1, respectively. Positive Control - WB: human placenta tissue, human U-87MG whole cell, human PC-3 whole cell, human MCF-7 whole cell, rat stomach tissue. ICC/IF: A549 cell. Flow: U87 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Neuropilin 1 is a 923 amino acid containing membrane bound coreceptor to a tyrosine kinase receptor belonging to the neuropilin family with two CUB domains, two F5/8 type C domains and a MAM domain. Neuropilin 1 forms a heterodimer with NRP2 (Neuropilin 2) and is known to bind to plexin (PLXNB1) to form semaphorin receptors. Expressed mostly in the nervous system this receptor is implicated in a wide range of functions including in the development of the cardiovascular system, angiogenesis, repulsive axon guidance, formation of certain neuronal circuits, cell survival, control of neuronal migration and organogenesis outside the nervous system. This semaphorin receptor functions as receptors for both the semaphorins and vascular endothelial growth factor and is also a regulator of VEGF-induced angiogenesis. Neuropilin 1 may be involved in aggressive advanced prostate carcinoma and in the progression of breast cancer.
Specifications
Neuropilin 1 | |
Monoclonal | |
500 μg/mL | |
PBS with 4mg trehalose and no preservative | |
O14786, Q9QWJ9 | |
NRP1 | |
E.coli-derived human Neuropilin 1 recombinant protein (Position: K504-T827). | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG2a |
Flow Cytometry, Western Blot, Immunocytochemistry | |
8G6G7 | |
Unconjugated | |
NRP1 | |
A5 protein; BDCA4; C530029I03; CD304; Neuropilin; neuropilin 1; Neuropilin1; neuropilin-1; Neuropilin-1 precursor (A5 protein); NP1; NP-1; Npn1; NPN-1; NRP; NRP 1; Nrp1; sNRP 1; sNRP1; soluble NRP 1; soluble NRP1; transmembrane receptor; vascular endothelial cell growth factor 165 receptor; VEGF165R | |
Mouse | |
Antigen affinity chromatography | |
RUO | |
246331, 8829 | |
-20°C | |
Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.