Learn More
Invitrogen™ Neuropilin-1 Recombinant Rabbit Monoclonal Antibody (9W1C10)

Description
Immunogen sequence: IDETGSTPGY EGEGEGDKNI SRKPGNVLKT LDPILITIIA MSALGVLLGA VCGVVLYCAC WHNGMSERNL SALENYNFEL VDGVKLKKDK LNTQSTYSEA.
Specifications
Specifications
| Antigen | Neuropilin-1 |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 9W1C10 |
| Concentration | 0.8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | NRP1 |
| Gene Accession No. | O14786, P97333, Q9QWJ9 |
| Gene Alias | A5 protein; BDCA4; C530029I03; CD304; Neuropilin; neuropilin 1; Neuropilin1; neuropilin-1; Neuropilin-1 precursor (A5 protein); NP1; NP-1; Npn1; NPN-1; NRP; NRP 1; Nrp1; sNRP 1; sNRP1; soluble NRP 1; soluble NRP1; transmembrane receptor; vascular endothelial cell growth factor 165 receptor; VEGF165R |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.