Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neuropilin-2 Antibody (96009R) [DyLight 650], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB567RW
Description
Neuropilin-2 Monoclonal antibody specifically detects Neuropilin-2 in Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, CyTOFSpecifications
Neuropilin-2 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Rat | |
Purified |
Western Blot, Simple Western, Flow Cytometry, CyTOF | |
96009R | |
Western Blot, Simple Western, Flow Cytometry, CyTOF-ready | |
MGC126574, neuropilin 2, neuropilin-2, neuropilin-2a(22), neuropilin-2b(0), NP2, NPN 2, NPN2, PRO2714, receptor for VEGF165 and semaphorins class3, Vascular endothelial cell growth factor 165 receptor 2, vascular endothelial growth factor-165 receptor 2, VEGF1265R2, VEGF165R2neuropilin-2a(17) | |
Mouse myeloma cell line NS0-derived recombinant rat Neuropilin-2, Gln23-Asp857 (Val809-Asp825 del), Accession # O35276 | |
0.1 mL | |
Angiogenesis, Cancer, Hypoxia | |
8828 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction