Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Neuropilin 2 Monoclonal Antibody (2A4H7C11)
GREENER_CHOICE

Catalog No. PIMA530698
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530698 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530698 Supplier Invitrogen™ Supplier No. MA530698
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human NRP2/Neurophilin-2.

This gene encodes a member of the neuropilin family of receptor proteins. The encoded transmembrane protein binds to SEMA3C protein (sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3C) and SEMA3F protein (sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3F), and interacts with vascular endothelial growth factor (VEGF). This protein may play a role in cardiovascular development, axon guidance, and tumorigenesis. Multiple transcript variants encoding distinct isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neuropilin 2
Applications ELISA
Classification Monoclonal
Clone 2A4H7C11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene NRP2
Gene Accession No. O60462
Gene Alias 1110048P06Rik; neuropilin 2; neuropilin-2; neuropilin-2a(17); neuropilin-2a(22); neuropilin-2b(0); Np2; Np-2; Npn2; Npn-2; NRP2; PRO2714; receptor for VEGF165 and semaphorins class3; vascular endothelial cell growth factor 165 receptor 2; VEGF165R2
Gene Symbols NRP2
Host Species Mouse
Immunogen Recombinant Human NRP2/Neurophilin-2 protein (Met1-Tyr855).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8828
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.