Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuropilin-2 Mouse anti-Human, Mouse, Rat, Alexa Fluor 750, Clone: 96009, R&D Systems™

Catalog No. FAB567S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB567S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB567S100 Supplier R&D Systems Supplier No. FAB567S100UG
This item is not returnable. View return policy

Mouse MonoclonalAntibody

Specifications

Antigen Neuropilin-2
Applications Flow Cytometry
Classification Monoclonal
Clone 96009
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. O35276
Gene Alias MGC126574, neuropilin 2, neuropilin-2, Neuropilin2, neuropilin-2a(22), neuropilin-2b(0), NP2, NPN 2, NPN2, PRO2714, receptor for VEGF165 and semaphorins class3, Vascular endothelial cell growth factor 165 receptor 2, vascular endothelial growth factor-165 receptor 2, VEGF1265R2, VEGF165R2neuropilin-2a(17)
Gene Symbols NRP2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant rat Neuropilin-2, Gln23-Asp857 (Val809-Asp825 del), Accession # O35276
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8828
Test Specificity Detects rat Neuropilin-2 in direct ELISAs and detects human, mouse, and rat Neuropilin-2 in Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human Neuropilin-1.
Target Species Human, Mouse, Rat
Content And Storage Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.