Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Neuropilin 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588742
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588742 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588742 Supplier Invitrogen™ Supplier No. PA588742
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GLIADSQISA SSTQEYLWSP SAARLVSSRS GWFPRIPQAQ PGEEWLQVDL GTPKTVKGVI IQGARGGDSI TAVEARAFVR KFKVSYSLNG KDWEYIQDPR TQQPKLFEGN MHYDTPDIRR FDPIPAQYVR VYPERWSPAG IGMRLEVLGC DWTDSKPTVE TLGPTVKSEE TTTPYPTEEE ATECGENCSF EDDKDLQLPS GFNCNFDFLE EPCGWMYDHA KWLRTTWASS SSPNDRTFPD DRNFLRLQSD SQREGQYARL ISPPVHLPRS PVCMEFQYQA TGGRGVALQV VREASQES; Positive Samples: Mouse kidney, Rat kidney; Cellular Location: Membrane, Single-pass type I membrane protein.

This gene encodes a member of the neuropilin family of receptor proteins. The encoded transmembrane protein binds to SEMA3C protein (sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3C) and SEMA3F protein (sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3F), and interacts with vascular endothelial growth factor (VEGF). This protein may play a role in cardiovascular development, axon guidance, and tumorigenesis. Multiple transcript variants encoding distinct isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neuropilin 2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.12 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NRP2
Gene Accession No. O35276, O35375
Gene Alias 1110048P06Rik; neuropilin 2; neuropilin-2; neuropilin-2a(17); neuropilin-2a(22); neuropilin-2b(0); Np2; Np-2; Npn2; Npn-2; NRP2; PRO2714; receptor for VEGF165 and semaphorins class3; vascular endothelial cell growth factor 165 receptor 2; VEGF165R2
Gene Symbols NRP2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 443-740 of human NRP2 (NP_0038632).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18187, 81527
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.