Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuropilin-2b Rabbit anti-Human, Mouse, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABN2262100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN2262-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN2262-100 Supplier MilliporeSigma Supplier No. ABN2262100UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Neuropilin-2b, Cat. No. ABN2262, is a highly specific rabbit polyclonal antibody that targets Neuropilin-2b (NRP2b) and has been tested for use in Immunocytochemistry,Immunohistochemistry, Immunoprecipitation, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neuropilin-2b
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Vascular endothelial cell growth factor 165 receptor 2;Neuropilin-2b.
Gene Symbols NRP2;VEGF165R2
Host Species Rabbit
Immunogen Mixture of two KLH-conjugated peptides corresponding to 17 and 16 amino acids from the C-terminal region of human Neuropilin-2b.
Molecular Weight of Antigen ∽115 kDa observed. 104.86 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_061004;NP_957719
Test Specificity This rabbit polyclonal antibody detects Neuropilin-2b in human and murine cells.
Target Species Human, Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.