Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neuroplastin Antibody (OTI3C4), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272966H
Description
Neuroplastin Monoclonal specifically detects Neuroplastin in Human, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Neuroplastin | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
NPTN | |
Full length recombinant protein of human Neuroplastin (NP_059429) produced in HEK293T cells. | |
0.1 mL | |
27020.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI3C4 | |
Western Blot, Immunohistochemistry | |
GP55Stromal cell-derived receptor 1, GP65SDR-1, MGC102805, neuroplastin, np55, np65, SDFR1DKFZp686L2477, SDR1stromal cell derived factor receptor 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Rat, Canine, Monkey | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction