Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neuroplastin Antibody (OTI3C4), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272966PE
Description
Neuroplastin Monoclonal antibody specifically detects Neuroplastin in Human, Rat, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
Neuroplastin | |
Monoclonal | |
PE | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
27020 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry | |
OTI3C4 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
GP55Stromal cell-derived receptor 1, GP65SDR-1, MGC102805, neuroplastin, np55, np65, SDFR1DKFZp686L2477, SDR1stromal cell derived factor receptor 1 | |
Full length human recombinant protein of human NPTN(NP_059429) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human, Rat, Canine, Monkey | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction