Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Neuroplastin Monoclonal Antibody (OTI3C4)
GREENER_CHOICE

Catalog No. PIMA525877
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525877 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525877 Supplier Invitrogen™ Supplier No. MA525877
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Neuroplastin Monoclonal Antibody for ICC/IF, IHC (P), Western Blot

This gene encodes a type I transmembrane protein belonging to the Ig superfamily. The protein is believed to be involved in cell-cell interactions or cell-substrate interactions. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neuroplastin
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI3C4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NPTN
Gene Accession No. P97546, Q9Y639
Gene Alias AW554172; glycoprotein 55; glycoprotein 55/65; glycoprotein 65; GP55; gp55/65; GP65; Neuroplastin; np55; np65; NPTN; Sdfr1; Sdr1; SDR-1; stromal cell derived factor receptor 1; stromal cell-derived receptor 1
Gene Symbols NPTN
Host Species Mouse
Immunogen Full length human recombinant protein of NPTN produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27020, 487637, 56064
Target Species Canine, Human, Monkey, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.