Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Neuroplastin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596882
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596882 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596882 Supplier Invitrogen™ Supplier No. PA596882
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QNEPRIVTSE EVIIRDSPVL PVTLQCNLTS SSHTLTYSYW TKNGVELSAT RKNASNMEYR INKPRAEDSG EYHCVYHFVS APKANATIEV KAAPDITGHK RSENKNEGQD ATMYCKSVGY PHPDWIWRKK ENGMPMDIVN TSGRFFIINK ENYTELNIVN LQITEDPGEY EC; Positive Samples: SKOV3, Jurkat; Cellular Location: Cell membrane, Single-pass type I membrane protein.

This gene encodes a type I transmembrane protein belonging to the Ig superfamily. The protein is believed to be involved in cell-cell interactions or cell-substrate interactions. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neuroplastin
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.79 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NPTN
Gene Accession No. P97300, P97546, Q9Y639
Gene Alias AW554172; glycoprotein 55; glycoprotein 55/65; glycoprotein 65; GP55; gp55/65; GP65; Neuroplastin; np55; np65; NPTN; Sdfr1; Sdr1; SDR-1; stromal cell derived factor receptor 1; stromal cell-derived receptor 1
Gene Symbols NPTN
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20320, 27020, 56064
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.