Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Neuroplastin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533987
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533987 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533987 Supplier Invitrogen™ Supplier No. PA533987
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Neuroplastin Polyclonal Antibody for IHC (P)

This gene encodes a type I transmembrane protein belonging to the Ig superfamily. The protein is believed to be involved in cell-cell interactions or cell-substrate interactions. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neuroplastin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene NPTN
Gene Accession No. P97300, P97546, Q9Y639
Gene Alias AW554172; glycoprotein 55; glycoprotein 55/65; glycoprotein 65; GP55; gp55/65; GP65; Neuroplastin; np55; np65; NPTN; Sdfr1; Sdr1; SDR-1; stromal cell derived factor receptor 1; stromal cell-derived receptor 1
Gene Symbols NPTN
Host Species Rabbit
Immunogen Synthetic 19 amino acid peptide from internal region of human NPTN.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100052106, 100155307, 100345982, 100720016, 100758174, 20320, 27020, 487637, 540039, 56064
Target Species Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Hamster, Pig, Horse, Guinea Pig, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.