Learn More
Stoelting™ NEUROS™ Syringes
SDP
Stoelting™ Hamilton Neuros™ syringe technology provides functionality for controlled animal injections.
$368.00 - $807.00
Specifications
For Use With (Application) Neuroscience
Model Blunt
View More Specs
Products 7
Catalog Number Mfr. No. Size Includes Price Quantity & Availability  
Catalog Number Mfr. No. Size Includes Price Quantity & Availability  
10-000-888
SDP
View Documents
Stoelting™
53430
100 μL, 33 gauge Adapter Kit
Each for $695.50
Only null left
Add to Cart
10-000-895
SDP
View Documents
Stoelting™
53437
5 to 100 μL, 33 gauge 6/pk.
Pack of 6 for $368.00
Only null left
Add to Cart
10-000-922
SDP
View Documents
Stoelting™
53495
2 μL, 30 gauge
Each for $807.00
Only null left
Add to Cart
10-000-923
SDP
View Documents
Stoelting™
53496
5 μL, 33 gauge Adapter Kit
Each for $694.50
Only null left
Add to Cart
10-000-924
SDP
View Documents
Stoelting™
53497
10 μL, 33 gauge Adapter Kit
Each for $642.00
Only null left
Add to Cart
10-000-925
SDP
View Documents
Stoelting™
53498
25 μL, 33 gauge Adapter Kit
Each for $695.50
Only null left
Add to Cart
10-000-926
SDP
View Documents
Stoelting™
53499
50 μL, 33 gauge Adapter Kit
Each for $674.50
Only null left
Add to Cart
Description
Description
  • Accurately dispenses 50 nL to 100 μL of liquid through an ultrafine needle protected by a custom-designed sleeve
  • Developed specifically for the neurosciences
  • Minimizes injection site damage by increasing needle rigidity and delivering a microvolume of liquid to an exact location
  • Flexible options for a variety of uses
  • Crafted to have very little to no dead volume which preserves the maximum amount of buffer and solution.
  • Blunt point (point style 3) ensures the sample is delivered directly to the site
  • Two different needle sleeves
  • Sleeve with a blind stop is ideal for cannulated applications and ensures targeted administration with an adjustable penetration depth
  • Version without a blind stop works best with stereotaxic holders
  • Both types provide an adjustable needle exposure of 0 to 20 mm
Specifications
Specifications
Neuroscience
Blunt
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.