Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neurotrimin/HNT Antibody (MM0485-5X25) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $579.29
Specifications
| Antigen | Neurotrimin/HNT |
|---|---|
| Clone | MM0485-5X25 |
| Concentration | LYOPH |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
Description
Neurotrimin/HNT Monoclonal specifically detects Neurotrimin/HNT in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Neurotrimin/HNT | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| Q9P121 | |
| 50863 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| MM0485-5X25 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| hNT, IgLON family member 2, IGLON2HNTNT, MGC60329, neurotrimin, NTRI | |
| NTM | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title