Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neurotrophin-4 Monoclonal Antibody (ANT-117), QED Bioscience™

Mouse Monoclonal Antibody
Supplier: Leinco Technologies Inc 501761000UG

This item is not returnable.
View return policy
Description
Neurotrophin-4 Monoclonal antibody specifically detects Neurotrophin-4 in Human samples. It is validated for ELISASpecifications
Neurotrophin-4 | |
Monoclonal | |
0.5-1.0 mg/mL | |
PBS with no preservative | |
P34130 | |
Mouse | |
Protein A | |
RUO | |
4909 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
ELISA | |
ANT-117 | |
Unconjugated | |
NTF4 | |
2900040K06Rik; AI462899; GLC10; GLC1O; neurotrophic factor 4; neurotrophic factor 5; neurotrophin 4; Neurotrophin 4/5; neurotrophin 5; Neurotrophin 5 (neurotrophin 4/5); neurotrophin 5 (neurotrophin 4/5); neurotrophin-4; Neurotrophin-5; neutrophic factor 4; Neutrophic factor 5; NT4; NT-4; NT4/5; NT-4/5; NT4/NT5; NT4P; NT5; NT-5; Ntf4; Ntf5; Ntf-5 | |
Recombinant human neurotrophin-4 | |
1 mg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction