Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neurotrypsin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP238231
Description
Neurotrypsin Polyclonal specifically detects Neurotrypsin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Neurotrypsin | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
P56730 | |
PRSS12 | |
This antibody was developed against a recombinant protein corresponding to amino acids: WVSVTDFGAPCLRWAEVPPFLERSPPASWAQLRGQRHNFCRSPDGAGRPWCFYGDARGKVDWGYCDCRHGSVRLRGGK | |
0.1 mL | |
Vision | |
8492 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
brain-specific serine protease 3, BSSP3, BSSP-3, EC 3.4.21, leydin, MGC12722, Motopsin, MRT1EC 3.4.21.-, neurotrypsin, protease, serine, 12 (neurotrypsin, motopsin), Serine protease 12 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction