Learn More
Description
Cortical neuron culture is an important model system for studies of neuronal development and function, neurotoxicity screening, drug discovery, and mechanisms of neurological diseases. Proper development and survival of neurons, in vivo as well as in vitro, requires specific growth factors, signaling molecules, peptides, and vitamins. The NeuroXVivo™ Rat Cortical Neuron Culture Kit provides optimized reagents and growth factors, as well as a validated protocol, to successfully culture rat cortical neurons in a serum-free environment. The kit contains a Neuronal Base Media, a neuronal media supplement, and growth factors to support the robust growth and maintenance of both short- and long-term neuronal cultures.
Applications: For culturing of either embryonic or postnatal cortical neurons
Storage: Store unopened product at -20 to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Specifications
Specifications
| Detection Method | Microscopy |
| Product Type | Neurologic Differentiation Kit |
| Quantity | 1 Kit |
| For Use With (Application) | Cell culture assays |
| Content And Storage | Neuronal Base Media, N21-MAX Media Supplement, Recombinant Human BDNF, Recombinant Human IGF-I, Reconstitution Buffer 1. Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
