Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neutec Group Accessories for Filling Systems
Shop All Neutec Group Products

Click to view available options
Product Type:
Double Tubing Set w/Filling Nozzle
F-Glass Piece
Filling Nozzle
Foot Switch
Petrirack
Pushout Table
Table
Tubing Set, Double
UV Lamp
Description
Filling nozzle with sliding tube for 8 x 12mm or 6 x 10mm tube
Specifications
Specifications
Product Type | Filling Nozzle |
For Use With (Equipment) | Neutec Group Media/Agar Filler PS 20 |
Includes | Sliding Tube |
Quantity | 1/Cs. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction