Learn More
Neutec Group Smart Dilutor

Shop All Neutec Group Products


Description
Includes
Both systems include polyethylene bag-holder, two sets of tubing, filters, 5L bottles, power cable, and an instruction manual. 14-910-315 is also supplied with two pumps.
Since these types of samples require a gravimetric type of dilution process, the Smart Dilutor has an integrated balance at the heart of the system. The operator simply places an unknown amount of sample in a sterile bag and the unit takes an initial weight of the sample, then the operator moves the lever and the microprocessor dispenses an amount of diluent by weight, to achieve the programmed dilution ratio. The system constantly monitors the weight in the sample bag to achieve a highly accurate dilution result. Once the programmed dilution ratio has been reached, the system notifies the operator that the process has finished.
- Software also allows the instrument to be used as a dispensing device for a previously determined quantity of a liquid into any kind of container
- Bar code provides traceability of the dilution process
- Can be expanded to dilute up to six different buffers with the use of an optional accessory
- Adjustable, multilevel arm enables dilution of various type containers
- Tubing set and the bottles are fully autoclavable
- Changing operation of tubing set is performed in less than 15 seconds
Certifications
Each device is calibrated and certified against equipment whose calibration is traceable to NIST™
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Product Type | Pipet Dispensing System |
Autoclavable | Autoclavable |
Dilution | 0.1 g |
Volume (Metric) | 1 μL to 25 mL |
For Use With (Application) | Dilution of solid, semi solid, or paste types of microbiological samples directly into sterile homogenizer bags |
Includes | Polyethylene bag holder, two sets of tubing, filters, 5L bottles, power cable, instruction manual, one pump |
Model | Single Pump |
Speed | 950 mL/min. |
Temperature (English) | 41°F to 104°F |
Temperature (Metric) | 5°C to 40°C |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.