Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
StatLab™ NeutraGreen™ Liquid Neutralizer



Click to view available options
Format:
Bottle
Cube
Packaging Quantity:
1 gal.
2.5 gal.
5 gal.
Description
- Comes in liquid form with no powders to mix or dissolve
- Waste reaction time of 2 to 3 hr enables same day disposal
- Third-party environmental data confirms NeutraGreen™ will neutralize waste formalin to a material that is non-toxic and non-hazardous
- Test with NeutraGreen™ Test Strips to ensure waste has been properly neutralized for drain disposal
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Capacity (English) | 1 gal. |
| Capacity (Metric) | 3.79 L |
| Material | Pre-treated Formalin Neutralizer |
| Type | Formalin Neutralizer |
| Format | Cube |
| For Use With (Application) | Neutralizing Waste Formalin |
| Packaging Quantity | 1 gal. |
| Weight (English) | 7.5 lb. |
| Weight (Metric) | 3.4 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction