Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Scigen VIP-Fixative Leak-Proof Specimen Containers Neutral Buffered 10% Formalin Filled
Shop All Scigen Inc Products
Click to view available options
Capacity (Metric):
120 mL, pre-filled 50%
180 mL, pre-filled 50%
1900 mL, pre-filled 50%
20 mL, pre-filled 50%
40 mL, pre-filled 50%
480 mL, pre-filled 50%
60 mL, pre-filled 50%
7500 mL, pre-filled 50%
90 mL, pre-filled 50%
960 mL, pre-filled 50%
Material:
HDPE
PET
Polypropylene
Color:
Clear
Translucent
White
Description
- Forma-Vial™ specimen containers are uniquely designed to be leak proof and protect against biohazardous or hazardous material release to the environment
- Highly visible OSHA compliant labeling
- Sizes range from 7mL dermatology containers to 7.5 Liters operating room organ buckets
- Container openings range from 13mm to 300mm
- Filled with 10% neutral buffered formalin to 50% of container’s capacity
- Formalin waste designed to be treated in bulk using Neutralex or treated individually using Neutralexr tablets (patent pending).
- Nonsterile containers marked with a lot code and expiration date, 4 year shelf life
- Manufactured using ISO 13485 and FDA quality systems
Specifications
Specifications
Product Type | Specimen Container Bucket |
Closure Included | Yes |
Graduated | No |
Material | HDPE |
Sterility | Non-sterile |
Capacity (English) | 2 gal., pre-filled 50% |
Capacity (Metric) | 7500 mL, pre-filled 50% |
Closure Color | White |
Closure Material | HDPE |
Closure Size | 292 mm |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction