Learn More
GVS™ Neutralizing Buffer Swabs 4mL
Catalog No. p-7144703 Shop All Osmonics Products
Change view
Click to view available options
Quantity:
125
500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-486-183 125
10-486-184 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-486-183 Supplier GVS™ Supplier No. 10498303
Add to Cart
Add to Cart
Neutralizing buffer swabs are used in the analysis of surfaces for total bacterial count. Permits transfer of swabbed organisms to the laboratory without loss in viability. Inactivates the bactericidal and bacteriostatic effects of chlorine and quaternary ammonium detergents.
Description
  • Neutralizing buffer inactivates the bactericidal and bacteriostatic effects of chlorine and quaternary ammonium detergents. However, neutralizing buffer exhibits no toxic effects on microorganisms. This permits the transfer of swabbed organisms to the laboratory without loss in viability.
Features and Benefits:
  • Individually wrapped
  • Qualitative and semi-quantitative hygiene control
  • Sterile packed and ready-for-use
  • Easy to handle
  • Rapid results
  • Long shelf-life
Applications:
  • Neutralizing buffer swabs are used in the monitoring of surfaces for total bacterial count.
  • Used for the collection of surface contamination from flat or convoluted surfaces prior to transport to a laboratory for culture and enumeration.
The SwabCheck principle
  • The surface is wiped with a cellulose swab and any bacteria collected are transferred via the swab into a tube containing a special medium with an indicator dye, which is then incubated.
  • A single bacterium is sufficient to cause a color change.
  • The SwabCheck is about 1000 times more sensitive than the conventional ATP method. This accuracy is particularly important in the food industry. With this simple method, it is possible to identify microorganisms such as Listeria monocytogenes, which must not be present in any concentration in food and beverages.
See 'GVS Life Sciences Microbiology Testing mpg YouTube' Video for further details on use.
Specifications
Applicator Material Cellulose
Sterility Sterile
Item Description Neutralizing buffer swabs for analysis of surfaces for total bacterial count.
Quantity 125
Neutralizing buffer is not designed to culture and enumerate microorganisms.