Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NeutrAvidin Protein, DyLight™ 594

Catalog No. PI22842
Click to view available options
Quantity:
1 mL

Protein

NeutrAvidin Protein, DyLight 594 conjugate, 22842, from Invitrogen., 22842, from Invitrogen. Species Reactivity: ; Applications: ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot.

Avidin is a glycoprotein found in the egg white and tissues of birds, reptiles and amphibia. The biotin-binding protein contains four identical subunits having a combined mass of 67,000-68,000 daltons. Removing the glycosylation from avidin yields NeutrAvidin Protein with a mass of 60,000 daltons. Carbohydrate-based lectin-binding is reduced to undetectable levels, yet biotin-binding affinity is retained. NeutrAvidin Protein offers the advantages of a neutral pI to minimize nonspecific adsorption, along with lysine residues that remain available for derivatization or other customized conjugation. NeutrAvidin Protein yields the lowest nonspecific binding among the known biotin binding proteins. The specific activity for biotin-binding is approximately 14ug/mg of protein, which is near the theoretical maximum activity. 22842 contains 1 mL of protein in magenta colored liquid at a concentration of 1mg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1mg/mL
Conjugate DyLight 594
Cross Reactivity Tag
Formulation PBS with 1% BSA and 0.02% sodium azide; pH 7.2
pH 7.2
Quantity 1 mL
Regulatory Status RUO
Content And Storage 4°C
Form Liquid
For Use With (Application) ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Purity or Quality Grade purified
Host Species N/A
Product Type NeutrAvidin
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.