Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neutrophil Elastase/ELA2 Antibody (6B6G6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261657
Description
Neutrophil Elastase/ELA2 Monoclonal specifically detects Neutrophil Elastase/ELA2 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
Neutrophil Elastase/ELA2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
Bone marrow serine protease, EC 3.4.21, EC 3.4.21.37, ELA2granulocyte-derived elastase, elastase 2, neutrophil, elastase, neutrophil expressed, Elastase-2, GE, HLEelastase-2, HNE, Human leukocyte elastase, leukocyte elastase, medullasin, NE, neutrophil elastase, PMN elastase, PMN-E, polymorphonuclear elastase, SCN1 | |
Mouse | |
28.5 kDa | |
0.1 mL | |
Primary | |
Human, Rat | |
Purified |
Western Blot, Flow Cytometry, ELISA | |
6B6G6 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
ELANE | |
Purified recombinant fragment of human ELANE (AA: 140-267) expressed in E. Coli. | |
Protein G purified | |
Cancer, Neuroscience | |
1991 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction