Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neutrophil Elastase/ELA2 Rabbit anti-Human, Mouse, Clone: 8X4E6, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$205.50 - $494.50

Specifications

Antigen Neutrophil Elastase/ELA2
Clone 8X4E6
Dilution Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB168540
SDP
View Documents
Novus Biologicals
NBP31672420UL
20 μg
Each for $205.50
Only null left
Add to Cart
 
NB168539
SDP
View Documents
Novus Biologicals
NBP316724100UL
100 μg
Each for $494.50
Only null left
Add to Cart
 
Description

Description

Neutrophil Elastase/ELA2 Monoclonal antibody specifically detects Neutrophil Elastase/ELA2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications

Specifications

Neutrophil Elastase/ELA2
Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Monoclonal
Purified
RUO
Human, Mouse
Bone marrow serine protease, EC 3.4.21, EC 3.4.21.37, ELA2granulocyte-derived elastase, elastase 2, neutrophil, elastase, neutrophil expressed, Elastase-2, GE, HLEelastase-2, HNE, Human leukocyte elastase, leukocyte elastase, medullasin, NE, neutrophil elastase, PMN elastase, PMN-E, polymorphonuclear elastase, SCN1
Recombinant fusion protein containing a sequence corresponding to amino acids 1041-1181 of human Neutrophil Elastase/ELA2 (P08246).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
8X4E6
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cancer, Neuroscience
PBS, 0.05% BSA, 50% glycerol, pH7.3
1991
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.