Learn More
Invitrogen™ Neutrophil elastase Monoclonal Antibody (39A)

Description
Mouse anti Human elastase antibody, clone 39A recognizes human neutrophil elastase, also known as elastase-2, Bone marrow serine protease, PMN elastase or Medullasin.
Specifications
Specifications
| Antigen | Neutrophil elastase |
| Applications | ELISA, Flow Cytometry, Functional Assay |
| Classification | Monoclonal |
| Clone | 39A |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with <0.1% sodium azide |
| Gene | ELANE |
| Gene Accession No. | P08246 |
| Gene Alias | bone marrow serine protease; ELA2; ELA2A; ELANE; Elastase; elastase 2, neutrophil; elastase, neutrophil expressed; Elastase2; elastase-2; F430011M15Rik; GE; granulocyte-derived elastase; HLE; HNE; Human leukocyte elastase; Leukocyte elastase; medullasin; NE; neutrophil elastase; PE-1; PMN elastase; PMN-E; polymorphonuclear elastase; SCN1; serine protease |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.